Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-BFP Rabbit Polyclonal Antibody

Supplier: BioVision

5994-100
10008-142EA 309 USD InStock
10008-142
Anti-BFP Rabbit Polyclonal Antibody
Antibodies
The Blue Fluorescent Protein (BFP) can be used as protein marker to study subcellular localization or translocation of proteins.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: BFP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: All Species

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty