Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-mCherry Rabbit Polyclonal Antibody

Supplier: BioVision

5993-100
10008-140EA 347.43 USD InStock
10008-140
Anti-mCherry Rabbit Polyclonal Antibody
Antibodies
mCherry is the second generation monomeric red fluorescent protein that have improved brightness and photostability. The recombinant mCherry is expressed and purified from transformed E. coli using a method that ensures high purity and maximal fluorescence intensity. mCherry protein is a 28.8 kDa monomer with 256 amino acid.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: mCherry
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: All Species

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty