Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GHRL Rabbit Polyclonal Antibody

Supplier: BioVision

5990-100
10008-128EA 252.35 USD InStock
10008-128
Anti-GHRL Rabbit Polyclonal Antibody
Antibodies
Ghrelin, a growth hormone (GH)-releasing acylated peptide, was identified as the endogenous ligand for the growth hormone (GH) secretagogue receptor 1a. It stimulates the release of GH from the anterior pituitary throμgh the GH secretagogue receptor (GHS-R). It may also play a role in regulating the digestive system and energy balance.

Recommended Dilutions: 0

Type: Primary
Antigen: Ghrelin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty