Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-JLP Rabbit Polyclonal Antibody

Supplier: BioVision

3736-100
10005-892EA 252.35 USD InStock
10005-892
Anti-JLP Rabbit Polyclonal Antibody
Antibodies
JLP (JNK-interacting leucine zipper protein) has been identified as a scaffolding protein that functions to position mitogen-activated protein kinases (MAPKs) into specific signaling modules so that successive phosphorylation events of the MAPK pathway can occur. JLP is also known as sperm associated antigen 9 (SPAG9) due to its abundant expression in testicular haploid germ cells.

Recommended Dilutions: Western Blot: 1:500-1:2500

Type: Primary
Antigen: JLP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty