Anti-Caper Rabbit Polyclonal Antibody

Supplier: BioVision
3733-100
10004-682EA 280.65 USD
10004-682
Anti-Caper Rabbit Polyclonal Antibody
Caper, also known as splicing factor HCC1 or hepatocellular carcinoma protein 1 and RNA binding region containing protein 2 (RNPC2), acts as a transcriptional coactivator for steroid nuclear receptors c-Jun, ERalpha and ER-beta. Caper, a nuclear protein with highest concentrations in nuclear speckles, plays a role in the pre-mRNA splicing process. Two isoforms of Caper, HCC1.3 and HCC1.4, co-localize with pre-mRNA splicing factor SC35 and uridine-rich small nuclear RNAs. Caper is a widely expressed protein with highest levels detected in skeletal muscle, lung, brain and pancreas.

Recommended Dilutions: Western Blot: 1:500-1:2500

Type: Primary
Antigen: Caper
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR