Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-KLK12 Rabbit Polyclonal Antibody

Supplier: BioVision

3720-100
10004-666EA 252.35 USD InStock
10004-666
Anti-KLK12 Rabbit Polyclonal Antibody
Antibodies
The human KLK gene family encodes about 15 serine proteases and has the potential to serve as cancer biomarkers. The Kallikrein 12 (KLK 12) is one the KLK members located on chromosome 19. Different splice variants of the KLK12 produces three transcript variants encoding different isoforms that can be found on amino acid 254, 248 and 110.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: KLK12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty