Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-VASP Rabbit Polyclonal Antibody

Supplier: BioVision

3718-100
10004-660EA 252.35 USD InStock
10004-660
Anti-VASP Rabbit Polyclonal Antibody
Antibodies
Vasodilator-stimulated phosphoprotein (VASP) belongs to the Ena/VASP family of adaptor proteins linking the cytoskeletal system to the signal transduction pathways. VASP functions in cytoskeletal organization, fibroblast migration, platelet activation and axon guidance. Three phosphorylation sites, Ser157, Ser239 and Thr278, have been identified. Evidence sμggests that VASP phosphorylation reduces its association with actin and has a negative effect on actin polymerization.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunohistochemistry: 5 µg/ml

Type: Primary
Antigen: VASP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty