Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-RBP4 Rabbit Polyclonal Antibody

Supplier: BioVision

3716-100
10004-658EA 272.95 USD InStock
10004-658
Anti-RBP4 Rabbit Polyclonal Antibody
Antibodies
RBP4 (Retinol Binding Protein 4). Retinol is transported in blood bound to the carrier protein RBP. Under physiological conditions, RBP binds transthyretin (TTR) to prevent glomerular filtration of low molecular weight RBP in the kidneys. The RBP/TTR molar ration provides an indirect way to indicate marginal vitamin A deficiency.

Recommended Dilutions: Western Blot: 1:500-1:1000

Type: Primary
Antigen: RBP4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty