Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-DMAP1 Rabbit Polyclonal Antibody

Supplier: BioVision

3715-100
10004-654EA 252.35 USD InStock
10004-654
Anti-DMAP1 Rabbit Polyclonal Antibody
Antibodies
DNA methylation can help regulate transcriptional silencing via repressive transcription complexes, which include methyl-CpG-binding domain protein and Histone deacetylases (HDACs) DNMT1. This core enzyme for mammalian DNA methylation can also establish a repressive transcription complex consisting of DNMT1, HDAC2 and DMAP1. DMAP1 can interact directly with the transcriptional corepressor TSG101. The DNMT1-DMAP1 exists throμghout the S-phase; HDAC2 joind DNMT1 and DMAP1 only during Late S phase. This provides a regulated means to deacetylate heterochromatin following replication.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: DMAP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty