Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-LRP5 + LRP6 Rabbit Polyclonal Antibody

Supplier: BioVision

3714-100
10004-650EA 252.35 USD InStock
10004-650
Anti-LRP5 + LRP6 Rabbit Polyclonal Antibody
Antibodies
LDL-related proteins LRP-5 and LRP-6. are members of the LDL receptor superfamily, which were found to be involved in the canonical Wnt signaling pathway. Mammalian LRP-6 was shown to bind to Wnt-1 and enhance Induced-induced developmental processes in Xenopus embryos. Mice lacking LRP-6 exhibited developmental defects that are similar to those caused by deficiencies in various Want proteins. Recent work also shows that Want proteins induce the binding of LRP-5 to Axin, leading Axin degradation and β-catenin stabilization.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: LRP5 + LRP6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty