Anti-SGLT1 Rabbit Polyclonal Antibody

Supplier: BioVision
AB97682-100
10004-642EA 321.33 USD
10004-642
Anti-SGLT1 Rabbit Polyclonal Antibody
Antibodies
Glucose transporters are integral membrane proteins that mediate the transport of glucose and structurally related substances across cellular membranes. Two families of glucose transporter have been identified: the facilitated diffusion glucose transporter family (GLUT family), also known as 'uniporters,' and the sodium dependent glucose transporter family (SGLT family), also known as 'cotransporters' or 'symporters'. The SGLT1 gene encodes a protein that is involved in the active transport of glucose and galactose into eukaryotic and some prokaryotic cells.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: SGLT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR