Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PLCG2 Rabbit Polyclonal Antibody

Supplier: BioVision

3710-100
10004-638EA 252.35 USD InStock
10004-638
Anti-PLCG2 Rabbit Polyclonal Antibody
Antibodies
PLC (Phosphoinositide-specific phospholipase C) plays a significant role in transmembrane signaling. Four members of PLCs have been identified: PLCβ, PLCg, PLCd, and PLCe. In response to extracellular stimuli (e.g., hormone, growth factors, neurotransmitters), PLC hydrolizes phosphatidylinositol 4,5-biphosphate (PIP2) into two secondary messengers: inositol 1,4,5-triphosphate (IP3) and diacylglycerol (DAG). PLCg2 is engaged in antigen-dependent signaling in B-cells and collagen-dependent signaling in platelets.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunohistochemistry: 5 µg/ml

Type: Primary
Antigen: PLCG2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty