Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-FOXP3 Rabbit Polyclonal Antibody

Supplier: BioVision

3709-100
10004-634EA 252.35 USD InStock
10004-634
Anti-FOXP3 Rabbit Polyclonal Antibody
Antibodies
FOXP3 a member of the forkhead/winge-helix family of transcriptional regulators and is highly conserved in humans. It has been proposed that FOXP3 may be a master regulatory gene and marker for Treg cells. Recent reports also sμggest expression of FOXP3 on subpopulation of rat CD8+ cells.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: FOXP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty