Anti-HAT-3 Rabbit Polyclonal Antibody

Supplier: BioVision
3707-100
10005-884EA 326.9 USD
10005-884
Anti-HAT-3 Rabbit Polyclonal Antibody
Histone acetyltransferases (HATs) have been implicated in a number of cellular functions including gene regulation, DNA synthesis, and repair. Histone acetyltransferases and deacetylases are respectively, the enzymes devoted to thr addition and removal of acetyl groups from lysine residues on the Histone N-terminal trails. The enzymes exert fundamental roles in developmental processes and their deregulation has been linked to the progression of diverse human disorders, including cancer.

Recommended Dilutions: Western Blot: 2-6 µg/ml

Type: Primary
Antigen: HAT-3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR