Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-KCNIP3 Rabbit Polyclonal Antibody

Supplier: BioVision

3706-100
10005-880EA 252.35 USD InStock
10005-880
Anti-KCNIP3 Rabbit Polyclonal Antibody
Antibodies
Calsenilin is also known as DREAM and KChlP3. Calsenilin interacts with presenilin-2, a protein involved in Alzheimer’s disease, to reverse presenilin mediated enhancement of calcium signaling. Calsenilin also has pro-apoptotic function, is a substrate for caspase-3, acts as a calcium regulated transcriptional suppressor, and interacts with A-type potassium channels.

Recommended Dilutions: Western Blot: 2-6 µg/ml

Type: Primary
Antigen: KCNIP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty