Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TUCAN/CARD8 Rabbit Polyclonal Antibody

Supplier: BioVision

3705-100
10005-876EA 252.35 USD InStock
10005-876
Anti-TUCAN/CARD8 Rabbit Polyclonal Antibody
Antibodies
TUCAN (Tumor Up-regulated Card-containing Antagonist of Caspase-Nine) is a CARD (Caspase-Associated Recruitment Domains) containing protein that binds and suppresses activation of pro-Caspase-9 and found to be overexpressed in some types of cancers. TUCAN binds specifically to pro-caspase-9 and interferes with its interaction with Apaf-1, thereby suppressing apoptosis signaling in the mitochondrial/cytochrome c pathway.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: TUCAN/CARD8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty