Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-BIF Rabbit Polyclonal Antibody

Supplier: BioVision

3704-100
10005-872EA 252.35 USD InStock
10005-872
Anti-BIF Rabbit Polyclonal Antibody
Antibodies
BIF-1 (Bax interacting factor-1) is a 365 amino acid protein that interacts with Bax protein and influences cell survival and death. BIF-1 contains a Src homology 3 (SH3) domain in the C-terminus. Overexpression of BIF-1 promotes Bax conformational change, caspase activation, and apoptotic cell death.

Recommended Dilutions: Western Blot: 2-6 µg/ml

Type: Primary
Antigen: BIF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty