Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-BIK Rabbit Polyclonal Antibody

Supplier: BioVision

3703-100
10005-868EA 252.35 USD InStock
10005-868
Anti-BIK Rabbit Polyclonal Antibody
Antibodies
BIK is a member of Pro-apoptotic Bcl-2 family proteins. Althoμgh Bik lacks Bcl-2 homologus region, BH1 and BH2, BIK interacts with Bcl-2 and Bcl-xL. BIK has been shown to induce apoptosis by activating caspase-9.

Recommended Dilutions: Western Blot: 2-6 µg/ml

Type: Primary
Antigen: BIK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty