Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-c-Jun Rabbit Polyclonal Antibody

Supplier: BioVision

3702-100
10005-602EA 272.95 USD InStock
10005-602
Anti-c-Jun Rabbit Polyclonal Antibody
Antibodies
Extracellular stimuli such as growth factors and UV irradiation stimulate phosphorylation of c-Jun at Ser63/73 and activate c-Jun dependent transcription. Mutation at Ser63/73 renders c-Jun nonresponsive to mitogenic and stress induced signaling pathways. JNK (c-Jun N-terminal Kinase) binds to the N-terminal region of c-Jun and phosphorylates c-Jun at Ser63/73. The activity of JNK is stimulated by the same signals that activate c-Jun.

Recommended Dilutions: Western Blot: 0.5-2 µg/ml; Immunohistochemistry: 20-40 µg/ml

Type: Primary
Antigen: c-Jun
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty