Anti-JNK1 Rabbit Polyclonal Antibody

Supplier: BioVision
3701-100
10005-598EA 286.04 USD
10005-598
Anti-JNK1 Rabbit Polyclonal Antibody
The JNK pathway is activated by largely distinct stimuli including inflammatory cytokines (such as TNF-α and IL-1), UV light, inhibitors of protein synthesis and osmotic stress. Activated MEKK1 phosphorylates SEK1 (also known as MKK4), which in turn activates SAPK (also known as JNK). JNK binds tightly to the N-terminal region of c-Jun and ATF-2, and phosphorylates c-Jun at Ser63 and Ser73 and ATF-2 at Thr69 and Thr71.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml;Immunoprecipitation: 10-20 µg/ml; and Immunohistochemistry: 10-20 µg/ml

Type: Primary
Antigen: JNK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR