Anti-Thromboxane B2 Rabbit Polyclonal Antibody

Supplier: BioVision
3700-100
10005-722EA 286.04 USD
10005-722
Anti-Thromboxane B2 Rabbit Polyclonal Antibody
Thromboxane B2 (TXB2) is further metabolized enzymatically to a series of compounds, of which 11-dehydro-TXB2 is the major product found both in plasma and urine. The dehydrogenase-catalyzing 11-dehydro-TXB2 formation is tissue bound and widely expressed, with the highest occurrence being found in lung and kidney.

Recommended Dilutions: Western Blot: 1-4 µg/ml

Type: Primary
Antigen: Thromboxane B2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR