Anti-XAF1 Rabbit Polyclonal Antibody

Supplier: BioVision
3696-100
10004-624EA 286.04 USD
10004-624
Anti-XAF1 Rabbit Polyclonal Antibody
XAF-1 binds to XIAP, an inhibitor of caspases-3, -7, -9 and triggers its relocation from the cytosol to the nucleus. Overexpression of XAF-1 results in the neutralization of XIAP’s ability to inhibit cell death. XAF-1is normally expressed in all adult and fetal tissues but was found to be present in very low levels in a variety of cancer cell lines. In contrast, XIAP levels have been shown to be high in a majority of cell lines.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: XAF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR