Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-BCL2L10 Rabbit Polyclonal Antibody

Supplier: BioVision

3695-100
10004-620EA 252.35 USD InStock
10004-620
Anti-BCL2L10 Rabbit Polyclonal Antibody
Antibodies
Bcl-B (also called Bcl2-L-10) is a novel anti-apoptotic member of the Bcl-2 family. Bcl-B is closest in amino acid sequence homology to Boo protein. It contains four Bcl-2 homologs. Bcl-B mRNA is widely expressed in adult human tissues. The Bcl-B protein binds to Bcl-2, Bcl-X(L), and Bax but not Bak. Bcl-B suppresses apoptosis induced by Bax, but not by Bak.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: BCL2L10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty