Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MAP1 Rabbit Polyclonal Antibody

Supplier: BioVision

3694-100
10004-616EA 252.35 USD InStock
10004-616
Anti-MAP1 Rabbit Polyclonal Antibody
Antibodies
Human MAP-1 (Modulator of Apoptosis Protein) is a novel Bax-associating protein. MAP-1 mediates caspase-dependent apoptosis in mammalian cells when overexpressed. MAP-1 homodimerizes and associates with the proapoptotic Bax and the prosurvival Bcl-2 and Bcl-XL of the Bcl-2 family.

Recommended Dilutions: Western Blot: 1-4 µg/ml

Type: Primary
Antigen: MAP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty