Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-NCS1 Rabbit Polyclonal Antibody

Supplier: BioVision

3693-100
10004-612EA 252.35 USD InStock
10004-612
Anti-NCS1 Rabbit Polyclonal Antibody
Antibodies
The neuronal calcium sensor (NCS) protein binds to calcium throμgh EF-hand motifs, are expressed in neurons. They are implicated in the regulation of neurotransmitters released at the neuromuscular junctions. NCS-1 is widely distributed in the brain, spinal cord and dorsal root ganglia. It has been shown to regulate neurosecreation in a calcium-dependent manner, potentiates nitric oxide synthase activity. NCS-1 has been reserved to indicate the protein observed in humans and mammals.

  • Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: NCS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty