Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HDC Rabbit Polyclonal Antibody

Supplier: BioVision

3691-100
10004-548EA 288.4 USD InStock
10004-548
Anti-HDC Rabbit Polyclonal Antibody
Antibodies
Histidine decarboxylase (HDC) is the enzyme catalyzing the conversion of histidine into histamine. HDC can be found in the histamine secreting ECL cells of some species as well as in the mast cells.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: HDC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty