Anti-HAT1 Rabbit Polyclonal Antibody

Supplier: BioVision
3689-100
10004-252EA 286.04 USD
10004-252
Anti-HAT1 Rabbit Polyclonal Antibody
Histone acetyltransferases (HATs) have been implicated in a number of cellular functions including gene regulation, DNA synthesis, and repair. Histone acetyltransferases and deacetylases are, respectively, the enzymes devoted to the addition and removal of acetyl groups from lysine residues on the histone N-terminal tails. The enzymes exert fundamental roles in developmental processes and their deregulation has been linked to the progression of diverse human disorders, including cancer.

Recommended Dilutions: Western Blot: 1-2 µg/ml

Type: Primary
Antigen: HAT-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR