Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HDAC3 Rabbit Polyclonal Antibody

Supplier: BioVision

3603-100
10004-354EA 252.35 USD InStock
10004-354
Anti-HDAC3 Rabbit Polyclonal Antibody
Antibodies
HDAC3 is a member of the class I mammalian histone deacetylase family, which plays an important role in modulating the eukaryotic chromatin structure. Human HDAC3 is composed of 428 amino acid residues. HDAC3 interacts with a growing number of transcriptional factors including SMRT and N-CoR. The interacting complexes bind to specific regions of chromatin and regulate gene transcription in these regions.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunohistochemistry: 20-30 µg/ml

Type: Primary
Antigen: HDAC3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Chicken

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty