Anti-HDAC2 Rabbit Polyclonal Antibody

Supplier: BioVision
3602-100
10004-350EA 285.6 USD
10004-350
Anti-HDAC2 Rabbit Polyclonal Antibody
HDAC2 is a class I mammalian histone deacetylase containing 488 amino acid residues with an apparent size of ~55 kDa by denaturing SDS-PAGE. HDAC2 has been shown to interact directly with transcription repressor complexes and the nuclear receptor corepressor N-CoR.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunocytochemistry: 10-20 µg/ml

Type: Primary
Antigen: HDAC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Chicken, Hamster, Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR