Anti-HDAC1 Rabbit Polyclonal Antibody

Supplier: BioVision
3601-100
10004-346EA 286.04 USD
10004-346
Anti-HDAC1 Rabbit Polyclonal Antibody
HDAC1 is an active component of transcriptional corepressor complexes which can be recruited to specific promoter regions via transcriptional factors. HDAC1 catalyzes removal of acetyl-groups from acetyl-lysines of histones and promotes compaction of chromatin in these regions, which leads to the inhibition of gene transcription.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunoprecipitation: 10-20 µg/ml; and Immunohistochemistry: 10-20 µg/ml

Type: Primary
Antigen: HDAC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR