Anti-CYP19A1 Rabbit Polyclonal Antibody

Supplier: BioVision
3599-100
10004-342EA 373.61 USD
10004-342
Anti-CYP19A1 Rabbit Polyclonal Antibody
Aromatase is a key enzyme in steroidogenesis and plays an important role in sexual differentiation, fertility, and carcinogenesis. Many environmental chemicals may influence aromatase activity and thereby disrupt endocrine function.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunohistochemistry: 10-20 µg/ml

Type: Primary
Antigen: CYP19A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR