Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-S6K1 Rabbit Polyclonal Antibody

Supplier: BioVision

3505-100
10004-160EA 370.8 USD InStock
10004-160
Anti-S6K1 Rabbit Polyclonal Antibody
Antibodies
p70 S6 kinase is a mitogen activated Ser/Thr protein kinase that is required for cell growth and G1 cell cycle progression. P70 S6 kinase phosphorylates the S6 protein of the 40S ribosomal subunit and is involved in translational control of 5’oligopyrimidine tract mRNAs. A second isoform, p85 S6 kinase, is derived from the same gene and is identical to p70 S6 kinase except for 23 extra residues at the N-terminus that encode a nuclear localizing signal. Both isoforms lie on a mitogen-activated signaling pathway downstream of phosphoinositide 3 kinase (PI3K) and the target of rapamycin FRAP/mTOR, and distinct from the Ras/MAP kinase cascade.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: S6K1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty