Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-RAF1 Rabbit Polyclonal Antibody

Supplier: BioVision

3504-100
10004-158EA 355.35 USD InStock
10004-158
Anti-RAF1 Rabbit Polyclonal Antibody
Antibodies
c-Raf is a mitogen-activated protein kinase. Binding of Raf to 14-3-3 is required for c-Raf kinase activity, and also blocks the ability of phosphatase to inhibit c-Raf. 14-3-3 binds c-Raf at two phospho-serine residues: Ser259 and Ser621. Mutation of Ser259 to alanine constitutively activates Raf kinase activity, but mutation of Ser621 to alanine results in an inactive kinase.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunohistochemistry: 20-40µg/ml

Type: Primary
Antigen: RAF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty