Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-JUN Rabbit Polyclonal Antibody

Supplier: BioVision

3502-100
10004-154EA 355.35 USD InStock
10004-154
Anti-JUN Rabbit Polyclonal Antibody
Antibodies
c-Jun is a component of the transcription factor AP-1 that binds and activates transcription at TRE/AP-1 elements. The transcriptional activity of c-Jun is regulated by phosphorylation at Ser63 and Ser73. Extracellular signals, such as growth factors and UV irradiation, stimulate phosphorylation of c-Jun at Ser63/73 and activate c-Jun dependent transcription. The MAP kinase homologue, SAPK/JNK, binds to the N-terminal region of c-Jun and phosphorylates c-Jun at Ser63/73.

Recommended Dilutions: Western Blot: 1 µg/ml; Immunohistochemistry: 10-20 µg/ml

Type: Primary
Antigen: JUN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty