Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-LCK Rabbit Polyclonal Antibody

Supplier: BioVision

3499-100
10004-152EA 355.35 USD InStock
10004-152
Anti-LCK Rabbit Polyclonal Antibody
Antibodies
Lck is a membrane associated tyrosine protein kinases. The Lck gene has been shown to undergo rearrangement and overexpression in some murine lymphomas. In human studies, it has been demonstrated that the Lck gene is localized to a site in the genome which undergoes frequent chromosomal abnormalities in lymphomas and neuroblastomas. Resting T cells contain high levels of the Lck protein and mRNA both of which decline upon activation of the T cells. These and other observations sμggest that alterations in Lck expression may contribute to the pathogenesis of some types of neoplastic disease.

Recommended Dilutions: Western Blot: 1-2 µg/ml; Immunoprecipitation: 10-20 µg/ml; and Immunohistochemistry: 20-25 µg/ml

Type: Primary
Antigen: LCK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty