Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HTRA2 Rabbit Polyclonal Antibody

Supplier: BioVision

3497R-100
10004-144EA 252.35 USD InStock
10004-144
Anti-HTRA2 Rabbit Polyclonal Antibody
Antibodies
Human HtrA2 (also designated Omi) is a novel member of the HtrA serine protease family and is highly homologous to HtrA (also known as L56 and HtrA1). HtrA2 is ubiquitously expressed nuclear protease that is capable of autoproteolysis. The HtrA2 protein exists as two polypeptides of 38 and 40 kDa and as an alternatively spliced form called D-Omi, which is predominately expressed in the kidney, colon and thyroid. Like HtrA, HtrA2 is involved in the degradation aberrantly folded proteins during conditions of cellular stress, sμggesting that it may possess a chaperone-like role under normal conditions.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: HTRA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty