Anti-Hsp47 Rabbit Polyclonal Antibody

Supplier: BioVision
AB18894-100
10004-092EA 347.56 USD
10004-092
Anti-Hsp47 Rabbit Polyclonal Antibody
Antibodies
Heat shock proteins (HSPs) are ubiquitously expressed in all organisms. HSP47 is a molecule expressed constitutively in cells that synthesizes collagen. HSP47 is a procollagen-specific molecular chaperon that resides in the endoplasmic reticulum of procollagen-producing cells and is essential for secretion of procollagen from cells. Significantly, HSP47 plays a vital role in folding and assembling collagen. HSP47 is also known as colligin 1, serpinh1, collagen-binding protein 1 (CBP1) and gp46. The human HSP 47 gene encodes a 47 kDa protein.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunoprecipitation: 10-20 µg/ml; Immunohistochemistry: 10-20 µg/ml (in frozen or paraffin embedded tissues)

Type: Primary
Antigen: Hsp47
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR