Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GSK3B Rabbit Polyclonal Antibody

Supplier: BioVision

3495-100
10004-090EA 355.35 USD InStock
10004-090
Anti-GSK3B Rabbit Polyclonal Antibody
Antibodies
Glycogen synthase kinase 3 (GSK-3) is a serine/threonine protein kinase that has been implicated in the regulation of cell fate and in the Wnt signaling pathway. GSK-3 plays an important role in the PI3 kinase and Akt mediated cell survival pathways, and its activity can be inhibited by Akt-mediated phosphorylation at Ser21 of GSK-3α and Ser9 of GSK-3β. GSK-3 has also been implicated in alzheimer’s disease. Six Tau protein isoforms have been identified, all of which are phosphorylated by GSK-3.

Recommended Dilutions: Western Blot: 1-2 µg/ml; Immunohistochemistry: 15-30 µg/ml

Type: Primary
Antigen: Phospho-GSK-3b
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty