Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-FIAF Rabbit Polyclonal Antibody

Supplier: BioVision

3490-100
10004-134EA 272.95 USD InStock
10004-134
Anti-FIAF Rabbit Polyclonal Antibody
Antibodies
Adipose tissue is an energy reserve in animals. Adipose cells produce and secrete numerous physiologically important proteins that help regulate whole body metabolism, such as adiponectin leptin. Fasting causes significant changes in nutrient metabolism and many of these changes are controlled by transcription factors that regulate rate-limiting enzymes. An important transcription factor that mediates metabolic processes induced by fasting is peroxisome proliferator-activated receptor alpha (PPAR alpha). PPAR alpha has recently been reported to target a novel gene encoding the secreted protein FIAF (fasting-induced adipose factor). FIAF is expressed predominantly in adipose tissue and is strongly upregulated in white adipose tissue and the liver under fasting conditions. Initial studies sμggest that FIAF represents a novel endocrine signal helping to regulate metabolism, especially under fasting conditions.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml;It recognizes ~50 kDa FIAF from samples of mouse and rat origins.

Type: Primary
Antigen: FIAF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty