Anti-EG-VEGF Goat Polyclonal Antibody

Supplier: BioVision
3489-100
10004-130EA 309.39 USD
10004-130
Anti-EG-VEGF Goat Polyclonal Antibody
Human EG-VEGF (Endocrine-gland-derived vascular endothelial growth factor) is a 9.6 kDa protein consisting of 86 amino acid residues. Human EG-VEGF induces proliferation, migration and fenestration in capillary endothelial cells derived from endocrine glands.

Recommended Dilutions: Western blotting at 1:1000 dilutions

Type: Primary
Antigen: EG-VEGF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR