Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CSTB Mouse Monoclonal Antibody [clone: RJMW2E7]

Supplier: BioVision

3487-100
10004-086EA 303.85 USD InStock
10004-086
Anti-CSTB Mouse Monoclonal Antibody [clone: RJMW2E7]
Antibodies
Cystatin B (stefin B, NCPI) is a natural inhibitor of lysosomal cysteine proteases (cathepsins). Its expression seems to be connected to the regulation of proteolysis which is essential for many physiological and pathological processes.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunohistochemistry: 10-20 µg/ml

Type: Primary
Antigen: CSTB
Clonality: Monoclonal
Clone: RJMW2E7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty