Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CSTA Mouse Monoclonal Antibody [clone: WR 23/2/3/3]

Supplier: BioVision

3486-100
10004-084EA 303.85 USD InStock
10004-084
Anti-CSTA Mouse Monoclonal Antibody [clone: WR 23/2/3/3]
Antibodies
Cystatin A (acid cysteine proteinase inhibitor, ACPI) is a natural inhibitor of cysteine proteinases and is located mainly in the keratohyaline granules of the stratum granulosum and the cornified envelope of the stratum corneum in the epidermis. Human cystatin A consists of 98 amino acid residues and belongs to cystatin subfamily I (stefins). Among the target enzymes inhibited by cystatin A are cathepsins B, C, H, L, and S, papain and actinidin. Cystatin A has also been detected in infiltrative breast carcinoma.

Recommended Dilutions: Western Blot: 1-4 µg/ml; Immunohistochemistry

Type: Primary
Antigen: CSTA
Clonality: Monoclonal
Clone: WR 23/2/3/3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty