Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-S6K1 Rabbit Polyclonal Antibody

Supplier: BioVision

3485-100
10004-082EA 272.95 USD InStock
10004-082
Anti-S6K1 Rabbit Polyclonal Antibody
Antibodies
p70 S6 kinase and p90 Rsk kinase are the two distinct 40S ribosomal protein S6 Ser/Thr kinases present in somatic animal cells. p90 Rsk kinase is maximally activated within minutes of addition of growth factors or phorbol ester to cultured cells followed by activation of p70 S6 kinase. Both enzymes are regulated by serine/threonine phosphorylation, sμggesting that specific kinases may exist upstream in the signaling pathways that regulate these kinases.

Recommended Dilutions: Western Blot: 1 µg/ml; Immunoprecipitation: 10-20 µg/ml; and Immunohistochemistry: 10-20 µg/ml

Type: Primary
Antigen: S6K1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty