Anti-PS2 Rabbit Polyclonal Antibody

Supplier: BioVision
3482-100
10004-074EA 309.39 USD
10004-074
Anti-PS2 Rabbit Polyclonal Antibody
Presenilin 1 and presenilin 2 are transmembrane proteins belonging to the presenilin family. Mutation of presenilin genes has been linked to early onset of Alzheimer’s disease, probably functioning throμgh its associated g-secretase activity for amyloid-β protein processing. Endogenous presenilin mainly exists in a heterodimeric complex formed from the endoproteolytically processed N-terminal (34 kDa) and C-terminal (~20 kDa) fragments.

Recommended Dilutions: Western Blot: 2-4 µg/ml; Immunoprecipitation: 20 µg/ml; and Immunohistochemistry: 20 µg/ml

Type: Primary
Antigen: PS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR