Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PSEN1 Rabbit Polyclonal Antibody

Supplier: BioVision

3481-100
10004-072EA 272.95 USD InStock
10004-072
Anti-PSEN1 Rabbit Polyclonal Antibody
Antibodies
Presenilin 1 and presenilin 2 are transmembrane proteins belonging to the presenilin family. Mutation of presenilin genes has been linked to early onset of Alzheimer’s disease, probably functioning throμgh its associated g-secretase activity for amyloid-β protein processing. Endogenous presenilin mainly exists in a heterodimeric complex formed from the endoproteolytically processed N-terminal (34 kDa) and C-terminal (~20 kDa) fragments.

Recommended Dilutions: Western Blot: 2-4 µg/ml

Type: Primary
Antigen: PSEN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty