Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PTEN Rabbit Polyclonal Antibody

Supplier: BioVision

3479-100
10004-022EA 272.95 USD InStock
10004-022
Anti-PTEN Rabbit Polyclonal Antibody
Antibodies
PTEN (phosphatase and tensin homologue deleted on chromosome ten) phosphatase is a tumor suppressor gene implicated in a wide variety of human cancers. PTEN encodes the 403 amino acid polypeptide originally described as a dual-specificity protein phosphatase. The main substrates of PTEN are inositol phospholipids generated by the activation of the phosphoinositide 3 kinase (PI3K). PTEN is a major negative regulator of the PI3K/Akt signaling pathway.

Recommended Dilutions: Western Blot: 1-2 µg/ml; Immunoprecipitation: 10-20 µg/ml

Type: Primary
Antigen: PTEN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty