Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-JKK1/SEK1/MKK4 Rabbit Polyclonal Antibody

Supplier: BioVision

3478-100
10004-020EA 334.75 USD InStock
10004-020
Anti-JKK1/SEK1/MKK4 Rabbit Polyclonal Antibody
Antibodies
SAPK/Erk kinase (SEK1), also known as MKK4 or Jun kinase kinase (JNKK), activates the MAP kinase homologues SAPK and JNK in response to various cellular stresses and inflammatory cytokines. Activation of SEK1 occurs throμgh phosphorylation of serine and threonine residues at positions 257 and 261, respectively, by MEKK. Like MEK, SEK is a dual-specificity protein kinase that phosphorylates SAPK/JNK at a conserved T*PY* site in its activation loop. Phosphorylation by Akt at Ser80 inhibits SEK1 and suppresses the stress-activated signal transduction.

Recommended Dilutions: Western Blot: 1-4 µg/ml

Type: Primary
Antigen: JKK1/SEK1/MKK4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty