Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SMAD5 Rabbit Polyclonal Antibody

Supplier: BioVision

3465-100
10005-648EA 272.95 USD InStock
10005-648
Anti-SMAD5 Rabbit Polyclonal Antibody
Antibodies
Smad proteins, the mammalian homologs of the Drosophila Mothers against dpp (Mad), have been implicated as downstream effectors of TGFβ/BMP signaling. Smad1, Smad5, and Smad8 are effectors of BMP2 and BMP4 function while Smad2 and Smad3 are involved in TGF-β and activin-mediated growth modulation. Smad4 has been shown to mediate all of the above activities throμgh interaction with various Smad family members. Smad6 and Smad7 regulate the response to activin/TGFβ signaling by interfering with TGFβ-mediated phosphorylation of other Smad family members.

Recommended Dilutions: Western Blot: 1-4 µg/ml

Type: Primary
Antigen: SMAD5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty