Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CASP9 Rabbit Polyclonal Antibody

Supplier: BioVision

3149-100
10013-872EA 442.55 USD InStock
10013-872
Anti-CASP9 Rabbit Polyclonal Antibody
Antibodies
Caspases are synthesized as inactive pro-enzymes that are processed to active form in cells undergoing apoptosis. Caspase-9 is an important member of the caspase family. Upon induction of apoptosis, Cytochrome c released from mitochondria associates with pro-caspase-9 (47 kDa) and Apaf-1. The complex processes pro-caspase-9 into a large subunit (37 kDa/17 kDa) and a small subunit (10 kDa). Cleaved caspase-9 further processes other caspases including caspase-3 and caspase-6, to initiate a caspase cascade leading to apoptosis. The affinity purified antibody recognizing the active forms of caspase-9 provides a new tool for identifying apoptotic cell populations in both tissue sections and cultured cells.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunoprecipitation: 10-20 µg/ml

Type: Primary
Antigen: CASP9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty