Anti-ARRB1 Rabbit Polyclonal Antibody

Supplier: BioVision
3148-100
10003-462EA 321.07 USD
10003-462
Anti-ARRB1 Rabbit Polyclonal Antibody
Beta arrestin 1 is a cofactor in BARK mediated beta adrenergic receptor desensitization. It also plays an important role in immune function which mediated by regulating receptor. Beta arrestin 1 is high expressed in nervous system, peripheral blood leukocytes.

Recommended Dilutions: Western Blot: 1-4 µg/ml

Type: Primary
Antigen: ARRB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR